Chroman-4-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chromanone

057978-25G 057978-100G
102518-870EA 186.15 USD
102518-870 102518-872
Chroman-4-one ≥95%
Chroman-4-one

Hazard: Irritant

Formula: C₉H₈O₂
MDL Number: MFCD00006840
CAS Number: 491-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR