2,6-Dibromo-4-nitropyridine ≥90%

Supplier: Matrix Scientific

057342-1G 057342-5G
102833-212EA 95.38 USD
102833-212 102833-214
2,6-Dibromo-4-nitropyridine ≥90%
2,6-Dibromo-4-nitropyridine

Hazard: Irritant

Formula: C₅H₂Br₂N₂O₂
MDL Number: MFCD07369947
CAS Number: 175422-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR