Lenalidomide

Supplier: Matrix Scientific

Synonyms: 3-(4-Amino-1-oxo-1,3-dihydro-2H-isoindol-2-yl)piperidine-2,6-dione, (R,S)-Lenalidomide, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione

064722-1G 064722-5G
102551-652EA 83.08 USD
102551-652 102551-654
Lenalidomide
Lenalidomide

Hazard: Irritant/Stored under Argon

Formula: C₁₃H₁₃N₃O₃
MW: 259.27 g/mol
Melting Pt: 269…271 °C
MDL Number: MFCD07772307
CAS Number: 191732-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR