2-Amino-6-methoxybenzothiazole

Supplier: Matrix Scientific

066397-25G 066397-100G
102833-412EA 23.08 USD
102833-412 102833-414
2-Amino-6-methoxybenzothiazole
2-Amino-6-methoxybenzothiazole

Hazard: Irritant

Formula: C₈H₈N₂OS
Melting Pt: 161…164 °C
MDL Number: MFCD00005787
CAS Number: 1747-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR