(1-Methylpyrrolidin-2-yl)methanol

Supplier: Matrix Scientific

Synonyms: 1-Methylpyrrolidine-2-methanol, (S)-(-)-2-Hydroxymethyl-1-methylpyrrolidine, 1-METHYL-2-PYRROLIDINEMETHANOL

065627-5G 065627-25G
102551-684EA 44.62 USD
102551-684 102551-686
(1-Methylpyrrolidin-2-yl)methanol
(1-Methylpyrrolidin-2-yl)methanol

Hazard: Irritant

Formula: C₆H₁₃NO
MW: 115.176 g/mol
MDL Number: MFCD00011727
CAS Number: 3554-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR