Ethyl isoxazole-4-carboxylate

Supplier: Matrix Scientific

065567-1G 065567-5G
102833-404EA 364.62 USD
102833-404 102833-406
Ethyl isoxazole-4-carboxylate
Ethyl isoxazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₃
MDL Number: MFCD07368244
CAS Number: 80370-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR