4-Methyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: Fomepizole

065514-5G 065514-25G
102551-680EA 44.62 USD
102551-680 102551-682
4-Methyl-1H-pyrazole ≥95%
4-Methyl-1H-pyrazole

Hazard: Irritant

Formula: C₄H₆N₂
MW: 82.11 g/mol
MDL Number: MFCD00005245
CAS Number: 7554-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR