2,6-Dichloro-4-nitroanisole
Supplier: Matrix Scientific
065429-5G
065429-25G
102833-396EA
195.38
USD
102833-396
102833-398
2,6-Dichloro-4-nitroanisole
2,6-Dichloro-4-nitroanisole
Hazard: Irritant
Formula:
C₇H₅Cl₂NO₃ |
MDL Number:
MFCD00061129 CAS Number: 17742-69-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...