2-Benzyl-1H-benzimidazole

Supplier: Matrix Scientific

Synonyms: 2-Benzyl-1H-benzo[d]imidazole

065354-1G 065354-5G
102833-382EA 15.38 USD
102833-382 102833-384
2-Benzyl-1H-benzimidazole
2-Benzyl-1H-benzimidazole

Hazard: Irritant

Formula: C₁₄H₁₂N₂
MW: 208.27 g/mol
MDL Number: MFCD00022680
CAS Number: 621-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR