L-Glutamic acid 5-benzyl ester

Supplier: Matrix Scientific

Synonyms: γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH

065148-25G 065148-100G
102551-674EA 106.15 USD
102551-674 102833-378
L-Glutamic acid 5-benzyl ester
L-Glutamic acid 5-benzyl ester

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Melting Pt: 162-165°C
MDL Number: MFCD00002633
CAS Number: 1676-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR