(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid
Supplier: Matrix Scientific
Synonyms:
Boc-Gln-OH, N(α)-Boc-L-glutamine, Nalpha-Boc-L-glutamine
065147-5G
065147-25G
102551-668EA
73.85
USD
102551-668
102551-670
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₈N₂O₅ MW: 246.3 g/mol |
MDL Number:
MFCD00065571 CAS Number: 13726-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...