2',4',6'-Trihydroxyacetophenone

Supplier: Matrix Scientific

Synonyms: Phloroacetophenone, THAP, 2-Acetylphloroglucinol

065026-25G 065026-100G
102551-656EA 52.31 USD
102551-656 102551-658
2',4',6'-Trihydroxyacetophenone
2',4',6'-Trihydroxyacetophenone

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 219…221 °C
MDL Number: MFCD00149091
CAS Number: 480-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR