Chrysin
Supplier: Matrix Scientific
Synonyms:
5,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxyflavone, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-phenyl-4-benzopyrone, 5,7-Dihydroxy-2-phenylchromen-4-one
064941-25G
064941-100G
102833-370EA
60
USD
102833-370
102833-372
Chrysin
Chrysin
Hazard: Irritant
Formula:
C₁₅H₁₀O₄ MW: 254.24 g/mol Boiling Pt: 429 Melting Pt: 284…289 °C |
MDL Number:
MFCD00006834 CAS Number: 480-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...