Thiazole-5-carboxyaldehyde

Supplier: Matrix Scientific

Synonyms: 5-Thiazolecarboxaldehyde, 5-Formylthiazole

064675-1G 064675-5G
102663-822EA 41.54 USD
102663-822 102663-824
Thiazole-5-carboxyaldehyde
Thiazole-5-carboxyaldehyde

Hazard: Irritant-Harmful/Refrigerate/Stored under Argon

Formula: C₄H₃NOS
Boiling Pt: 92…94 °C (16 mmHg)
Density: 1.304
Flash Pt: 98 °C
MDL Number: MFCD02179516
CAS Number: 1003-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR