3,3'-Dichlorobenzidine

Supplier: Matrix Scientific

063869-5G 063869-25G
102833-340EA 3023.08 USD
102833-340 102833-342
3,3'-Dichlorobenzidine
3,3'-Dichlorobenzidine

TCA listed: T
Hazard: Irritant

Formula: C₁₂H₁₀Cl₂N₂
Boiling Pt: 402°C
Melting Pt: 132-133°C
CAS Number: 91-94-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR