3-Hydroxy-p-anisic acid

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-4-methoxybenzoic acid, Isovanillic acid

063756-25G 063756-100G
102551-636EA 110.77 USD
102551-636 102551-638
3-Hydroxy-p-anisic acid
3-Hydroxy-p-anisic acid

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327
MDL Number: MFCD00002507
CAS Number: 645-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR