1-Adamantanemethanol

Supplier: Matrix Scientific

Synonyms: 1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol

063693-5G 063693-100G
102833-334EA 18.46 USD
102833-334 102833-338
1-Adamantanemethanol
1-Adamantanemethanol

Hazard: Irritant

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
MDL Number: MFCD00074751
CAS Number: 770-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR