5-Methoxy-2-nitrophenol ≥95%

Supplier: Matrix Scientific

063641-1G 063641-5G 063641-25G
102551-626EA 306.15 USD
102551-626 102551-628 102551-630
5-Methoxy-2-nitrophenol ≥95%
5-Methoxy-2-nitrophenol

Hazard: Irritant

Formula: C₇H₇NO₄
MDL Number: MFCD00100932
CAS Number: 704-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR