5-Thiazolecarboxylic acid
Supplier: Matrix Scientific
063587-1G
063587-5G
102551-614EA
33.85
USD
102551-614
102551-616
5-Thiazolecarboxylic acid
5-Thiazolecarboxylic acid
Hazard: Irritant/Refrigerate
Formula:
C₄H₃NO₂S Boiling Pt: 107-108°C/10 Torr Melting Pt: 196-197°C |
MDL Number:
MFCD03428539 CAS Number: 14527-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...