3,5-Dimethyl-1,2,4-triazole

Supplier: Matrix Scientific

Synonyms: 3,5-dimethyl-4H-1,2,4-triazole

063455-1G 063455-5G
102663-808EA 41.54 USD
102663-808 102663-810
3,5-Dimethyl-1,2,4-triazole
3,5-Dimethyl-1,2,4-triazole

Hazard: Irritant

Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00656686
CAS Number: 7343-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR