5-Methyl-1H-tetrazole
Supplier: Matrix Scientific
063453-5G
063453-25G
102551-606EA
76.92
USD
102551-606
102551-608
5-Methyl-1H-tetrazole
5-Methyl-1H-tetrazole
Hazard: Irritant
Formula:
C₂H₄N₄ MW: 84.08 g/mol |
MDL Number:
MFCD00185824 CAS Number: 4076-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...