trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-)

Supplier: Matrix Scientific

Synonyms: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

063430-25G 063430-100G
102551-596EA 166.15 USD
102551-596 102551-598
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-)
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
Melting Pt: 169…173 °C
MDL Number: MFCD00004400
CAS Number: 537-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR