4-Iodo-3-methyl-1H-pyrazole
Supplier: Matrix Scientific
063354-1G
063354-5G
102518-894EA
38.46
USD
102518-894
102518-896
4-Iodo-3-methyl-1H-pyrazole
4-Iodo-3-methyl-1H-pyrazole
Hazard: Irritant
Formula:
C₄H₅IN₂ |
MDL Number:
MFCD09908176 CAS Number: 15802-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...