2-Amino-6-methylphenol ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Amino-o-cresol, 6-Amino-2-methylphenol
063143-5G
063143-25G
102833-328EA
140
USD
102833-328
102833-330
2-Amino-6-methylphenol ≥95%
2-Amino-6-methylphenol
Hazard: Irritant
Formula:
C₇H₉NO MW: 123.1525 g/mol |
MDL Number:
MFCD03788432 CAS Number: 17672-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...