α-Bromo-p-toluonitrile

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)benzonitrile, 4-Cyanobenzyl bromide, p-(Bromomethyl)benzonitrile

062621-25G 062621-100G
102551-588EA 64.62 USD
102551-588 102551-590
α-Bromo-p-toluonitrile
α-Bromo-p-toluonitrile

Hazard: Toxic/Corrosive/Lachrymator

Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 113-117°C
MDL Number: MFCD00001829
CAS Number: 17201-43-3
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR