2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid

Supplier: Matrix Scientific

062509-1G 062509-5G 062509-25G
102663-794EA 49.23 USD
102663-794 102663-796 102663-798
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid

Hazard: Irritant

Formula: C₇H₂F₄O₃
MW: 210.08
Melting Pt: 148…150 °C (dec.)
MDL Number: MFCD00129952
CAS Number: 652-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR