4-Bromo-N,N-dimethylbenzamide

Supplier: Matrix Scientific

062491-1G 062491-5G
102833-316EA 86.15 USD
102833-316 102833-318
4-Bromo-N,N-dimethylbenzamide
4-Bromo-N,N-dimethylbenzamide

Hazard: Irritant

Formula: C₉H₁₀BrNO
MW: 228.0900 g/mol
MDL Number: MFCD00463696
CAS Number: 18469-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR