6-Quinolineboronic acid

Supplier: Matrix Scientific

Synonyms: Quinolin-6-ylboronic acid, Quinoline-6-boronic acid

062386-1G 062386-5G
102551-582EA 76.92 USD
102551-582 102551-584
6-Quinolineboronic acid
6-Quinolineboronic acid

Hazard: Irritant

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: 106…109 °C
MDL Number: MFCD03093100
CAS Number: 376581-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR