4,5-Dihydro-6-methyl-3(2H)-pyridazinone

Supplier: Matrix Scientific

Synonyms: 6-Methyl-4,5-dihydropyridazin-3(2H)-one

062359-5G 062359-25G
102551-578EA 76.92 USD
102551-578 102551-580
4,5-Dihydro-6-methyl-3(2H)-pyridazinone
4,5-Dihydro-6-methyl-3(2H)-pyridazinone

Hazard: Irritant

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Boiling Pt: 265-270°C
Melting Pt: 105 °C
MDL Number: MFCD00006455
CAS Number: 5157-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR