1,1,1-Trifluoropropan-2-amine hydrochloride

Supplier: Matrix Scientific

Synonyms: 1,1,1-Trifluoropropan-2-amine hydrochloride, 2-Amino-1,1,1-trifluoropropane hydrochloride

061036-1G 061036-5G
102663-788EA 250.77 USD
102663-788 102663-790
1,1,1-Trifluoropropan-2-amine hydrochloride
1,1,1-Trifluoropropan-2-amine hydrochloride

Hazard: Irritant

Formula: C₃H₆F₃N·ClH
MW: 149.543 g/mol
MDL Number: MFCD04972986
CAS Number: 2968-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR