2-(2-Bromoethyl)pyridine

Supplier: Matrix Scientific

Synonyms: 2-Bromoethyl-pyridine

060926-1G 060926-5G
102833-304EA 481.54 USD
102833-304 102833-306
2-(2-Bromoethyl)pyridine
2-(2-Bromoethyl)pyridine

Hazard: Irritant

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD04038851
CAS Number: 39232-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR