Methyl-4-aminobutanoate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-γ-Abu-OMe.HCl

060909-5G 060909-25G
102551-568EA 27.69 USD
102551-568 102551-570
Methyl-4-aminobutanoate hydrochloride ≥95%
Methyl-4-aminobutanoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₁NO₂·HCl
MW: 153.61 g/mol
Melting Pt: 120…125 °C
MDL Number: MFCD00043270
CAS Number: 13031-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR