2-Bromophenylhydrazinium chloride
Supplier: Matrix Scientific
Synonyms:
2-Bromophenylhydrazine hydrochloride, 1-(2-Bromophenyl)hydrazine hydrochloride
060603-25G
060603-100G
102663-782EA
84.62
USD
102663-782
102663-784
2-Bromophenylhydrazinium chloride
2-Bromophenylhydrazinium chloride
Hazard: Irritant
Formula:
C₆H₈BrClN₂ MW: 223.5 g/mol |
MDL Number:
MFCD00012926 CAS Number: 50709-33-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...