Ethyl-4-pyrazolecarboxylate

Supplier: Matrix Scientific

Synonyms: ethyl 1H-pyrazole-4-carboxylate, Ethyl 4-pyrazolecarboxylate

060516-5G 060516-25G
102663-778EA 1267.69 USD
102663-778 102663-780
Ethyl-4-pyrazolecarboxylate
Ethyl-4-pyrazolecarboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂
MDL Number: MFCD00010844
CAS Number: 37622-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR