N1-Methylbenzene-1,2-diamine dihydrochloride
Supplier: Matrix Scientific
Synonyms:
N-Methyl-1,2-benzenediamine dihydrochloride, N-Methyl-1,2-phenylenediamine dihydrochloride
060353-25G
060353-100G
102551-562EA
41.54
USD
102551-562
102551-564
N1-Methylbenzene-1,2-diamine dihydrochloride
N1-Methylbenzene-1,2-diamine dihydrochloride
Hazard: Irritant
Formula:
C₇H₁₂Cl₂N₂ MW: 195.0900 g/mol Melting Pt: 192 °C |
MDL Number:
MFCD00042021 CAS Number: 25148-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...