Ethyl-4-(3-ethoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥99%

Supplier: Matrix Scientific

060056-1G 060056-5G
102518-882EA 286.15 USD
102518-882 102518-884
Ethyl-4-(3-ethoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥99%
Ethyl-4-(3-ethoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₁NO₄
MDL Number: MFCD00010644
CAS Number: 54278-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR