5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine
Supplier: Matrix Scientific
059426-1G
059426-5G
102833-280EA
90.77
USD
102833-280
102833-282
5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine
5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine
Hazard: Irritant
Formula:
C₆H₃BrIN₃ MW: 323.9166 g/mol |
MDL Number:
MFCD09878697 CAS Number: 875781-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...