Ethyl-2-ethylacetoacetate
Supplier: Matrix Scientific
058475-25G
058475-100G
102551-538EA
138.46
USD
102551-538
102551-540
Ethyl-2-ethylacetoacetate
Ethyl-2-ethylacetoacetate
Formula:
C₈H₁₄O₃ |
MDL Number:
MFCD00039898 CAS Number: 607-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...