1-Phenyl-1H-pyrazole

Supplier: Matrix Scientific

058385-25G 058385-100G
102551-530EA 98.46 USD
102551-530 102551-532
1-Phenyl-1H-pyrazole
1-Phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₉H₈N₂
MDL Number: MFCD00003112
CAS Number: 1126-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR