Methyl imidothiocarbamate sulfate

Supplier: Matrix Scientific

058383-25G 058383-100G
102833-264EA 112.31 USD
102833-264 102833-266
Methyl imidothiocarbamate sulfate
Methyl imidothiocarbamate sulfate

Hazard: Irritant

Formula: C₂H₈N₂O₄S₂
MDL Number: MFCD00135509
CAS Number: 14527-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR