N,N-Dimethylacetamide dimethyl acetal ≥85%
Supplier: Matrix Scientific
Synonyms:
(1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine
058342-5G
058342-25G
102551-526EA
26.15
USD
102551-526
102551-528
N,N-Dimethylacetamide dimethyl acetal ≥85%
N,N-Dimethylacetamide dimethyl acetal
Hazard: Flammable/Irritant
Formula:
C₆H₁₅NO₂ Boiling Pt: 118 °C (760 mmHg) Density: 0.911 Flash Pt: 8 °C |
MDL Number:
MFCD00008476 CAS Number: 18871-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...