6-Iodopyridazin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-6-iodopyridazine

058078-1G 058078-5G
102518-874EA 60 USD
102518-874 102518-876
6-Iodopyridazin-3-amine ≥95%
6-Iodopyridazin-3-amine

Hazard: Irritant

Formula: C₄H₄IN₃
MW: 221 g/mol
Boiling Pt: 399 °C
Melting Pt: 158…162 °C
Density: 2.204
Flash Pt: 195 °C
MDL Number: MFCD09991754
CAS Number: 187973-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR