6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Paliperidone Related Compound B
057886-5G
057886-25G
102833-242EA
104.62
USD
102833-242
102833-244
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride ≥95%
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₄ClFN₂O MW: 256.7000 g/mol |
MDL Number:
MFCD06658529 CAS Number: 84163-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...