4-Hydroxy-2,6-dimethylbenzoic acid ≥95%

Supplier: Matrix Scientific

057729-1G 057729-5G
102518-860EA 138.46 USD
102518-860 102518-862
4-Hydroxy-2,6-dimethylbenzoic acid ≥95%
4-Hydroxy-2,6-dimethylbenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
Melting Pt: 178 °C
MDL Number: MFCD00963146
CAS Number: 75056-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR