4-Phenylbenzyl bromide ≥96%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl

057609-5G 057609-25G
102551-486EA 73.85 USD
102551-486 102551-488
4-Phenylbenzyl bromide ≥96%
4-Phenylbenzyl bromide

Hazard: Irritant

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
MDL Number: MFCD00017869
CAS Number: 2567-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR