2-Bromo-5-fluoro-4-methylpyridine ≥95%
Supplier: Matrix Scientific
057409-1G
057409-5G
102518-846EA
60
USD
102518-846
102518-848
2-Bromo-5-fluoro-4-methylpyridine ≥95%
2-Bromo-5-fluoro-4-methylpyridine
Hazard: Irritant
Formula:
C₆H₅BrFN MW: 190.02 g/mol |
MDL Number:
MFCD08277310 CAS Number: 885168-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...