2-Methoxy-4-methyl-5-nitropyridine ≥95%
Supplier: Matrix Scientific
057341-5G
057341-25G
102833-208EA
40
USD
102833-208
102833-210
2-Methoxy-4-methyl-5-nitropyridine ≥95%
2-Methoxy-4-methyl-5-nitropyridine
Hazard: Irritant
Formula:
C₇H₈N₂O₃ |
MDL Number:
MFCD03095075 CAS Number: 6635-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...