4-(Trifluoromethyl)hydrocinnamic acid
Supplier: Matrix Scientific
Synonyms:
3-[4-(Trifluoromethyl)phenyl]propanoic acid
057201-5G
057201-25G
102551-446EA
32.31
USD
102551-446
102551-448
4-(Trifluoromethyl)hydrocinnamic acid
4-(Trifluoromethyl)hydrocinnamic acid
Hazard: Irritant
Formula:
C₁₀H₉F₃O₂ MW: 218.17 g/mol Melting Pt: 110…114 °C |
MDL Number:
MFCD00674032 CAS Number: 53473-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...