(4-Hydroxyphenyl)acetic acid ethyl ester ≥97%

Supplier: Matrix Scientific

057072-25G 057072-100G
102551-438EA 98.46 USD
102551-438 102551-440
(4-Hydroxyphenyl)acetic acid ethyl ester ≥97%
(4-Hydroxyphenyl)acetic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₂O₃
Boiling Pt: 110 °C (1 mmHg)
Melting Pt: 38…40 °C
MDL Number: MFCD00016491
CAS Number: 17138-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR