3-Bromo-4-nitrobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-nitro-4-(trifluoromethyl)benzene

054950-1G 054950-5G 054950-25G
102663-742EA 136.92 USD
102663-742 102663-744 102663-746
3-Bromo-4-nitrobenzotrifluoride ≥95%
3-Bromo-4-nitrobenzotrifluoride

Hazard: Irritant

Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
MDL Number: MFCD09998160
CAS Number: 132839-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR